Learn More
Bioss KD-Validated Chromosome 3 open
SDP
Catalog No. 5000065167
Encompass_Preferred
Chromosome 3 Open Reading Frame 38 Primary Antibody Unconjugated Public Immunogen Range 51-200/329 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65167 Supplier Bioss Supplier No. BSM90063R
Chromosome 3 Open Reading Frame 38 Primary Antibody Unconjugated Public Immunogen Range 51-200/329 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide