Learn More
Bioss KD-Validated Citrate synthetas
SDP
Catalog No. 5000064999
Encompass_Preferred
Citrate Synthase Primary Antibody Unconjugated Public Immunogen Range 400-466/466 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64999 Supplier Bioss Supplier No. BSM90363R
Citrate Synthase Primary Antibody Unconjugated Public Immunogen Range 400-466/466 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide