Learn More
Bioss KD-Validated cleaved Caspase-9
SDP
Catalog No. 5000065211
Encompass_Preferred
Caspase 9 Primary Antibody Unconjugated Public Immunogen Range 90-350/416 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65211 Supplier Bioss Supplier No. BSM90174R
Caspase 9 Primary Antibody Unconjugated Public Immunogen Range 90-350/416 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide