Learn More
Bioss KD-Validated CLIP1 Rabbit Reco
SDP
Catalog No. 5000065890
Encompass_Preferred
CAP-Gly Domain Containing Linker Protein 1 Primary Antibody Unconjugated Public Immunogen Range 1400-1438/1438 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65890 Supplier Bioss Supplier No. BSM90056R
CAP-Gly Domain Containing Linker Protein 1 Primary Antibody Unconjugated Public Immunogen Range 1400-1438/1438 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide