Learn More
Bioss KD-Validated Complement C3 Rab
SDP
Catalog No. 5000066834
Encompass_Preferred
Complement C3 Primary Antibody Unconjugated Public Immunogen Range 850-900/1663 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66834 Supplier Bioss Supplier No. BSM90042R
Complement C3 Primary Antibody Unconjugated Public Immunogen Range 850-900/1663 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide