Learn More
ABclonal Technology KD Validated COPB2 Rabbit
SDP
Catalog No. 5000007186
Encompass_Preferred
The Golgi coatomer complex (see MIM 601924) constitutes the coat of nonclathrin-coated vesicles and is essential for Golgi budding and vesicular trafficking It consists of 7 protein subunits including COPB2
Catalog No. 50-000-07186 Supplier ABclonal Technology Supplier No. A22646PM20UL
The Golgi coatomer complex (see MIM 601924) constitutes the coat of nonclathrin-coated vesicles and is essential for Golgi budding and vesicular trafficking It consists of 7 protein subunits including COPB2