Learn More
Bioss KD-Validated Core-binding fact
SDP
Catalog No. 5000065006
Encompass_Preferred
Core-Binding Factor Subunit Beta Primary Antibody Unconjugated Public Immunogen Range 100-182/182 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65006 Supplier Bioss Supplier No. BSM90099R
Core-Binding Factor Subunit Beta Primary Antibody Unconjugated Public Immunogen Range 100-182/182 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide