Learn More
Bioss KD-Validated CRKII Rabbit Reco
SDP
Catalog No. 5000066210
Encompass_Preferred
Adapter molecule crk Primary Antibody Unconjugated Public Immunogen Range 1-50/304 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66210 Supplier Bioss Supplier No. BSM90356R
Adapter molecule crk Primary Antibody Unconjugated Public Immunogen Range 1-50/304 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide