Learn More
Bioss KD-Validated CTP synthase 1 Ra
SDP
Catalog No. 5000064718
Encompass_Preferred
CTP Synthase 1 Primary Antibody Unconjugated Public Immunogen Range 250-300/591 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64718 Supplier Bioss Supplier No. BSM90188R
CTP Synthase 1 Primary Antibody Unconjugated Public Immunogen Range 250-300/591 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide