Learn More
ABclonal Technology KD Validated Cyclophilin F
SDP
Catalog No. 5000011642
Encompass_Preferred
The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins This protein is part of the mitochondrial permeability transition pore in the inner mitochondrial membrane Activation of this pore is thought to be involved in the induction of apoptotic and necrotic cell death
Catalog No. 50-000-11642 Supplier ABclonal Technology Supplier No. A25355100UL
The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins This protein is part of the mitochondrial permeability transition pore in the inner mitochondrial membrane Activation of this pore is thought to be involved in the induction of apoptotic and necrotic cell death