Learn More
Bioss KD-Validated Cystathionine bet
SDP
Catalog No. 5000065003
Encompass_Preferred
Cystathionine Beta-Synthase Primary Antibody Unconjugated Public Immunogen Range 1-69/654 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65003 Supplier Bioss Supplier No. BSM90403R
Cystathionine Beta-Synthase Primary Antibody Unconjugated Public Immunogen Range 1-69/654 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide