Learn More
Bioss KD-Validated Cytoplasmic linke
SDP
Catalog No. 5000066953
Encompass_Preferred
Cytoplasmic Linker Associated Protein 1 Primary Antibody Unconjugated Public Immunogen Range 1-52/1538 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66953 Supplier Bioss Supplier No. BSM90166R
Cytoplasmic Linker Associated Protein 1 Primary Antibody Unconjugated Public Immunogen Range 1-52/1538 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide