Learn More
Bioss KD-Validated DEK Rabbit Recomb
SDP
Catalog No. 5000066639
Encompass_Preferred
DEK Proto-Oncogene Primary Antibody Unconjugated Public Immunogen Range 405-455/1050 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66639 Supplier Bioss Supplier No. BSM90408R
DEK Proto-Oncogene Primary Antibody Unconjugated Public Immunogen Range 405-455/1050 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide