Learn More
Bioss KD-Validated Desmocollin 1 Rab
SDP
Catalog No. 5000066807
Encompass_Preferred
Desmocollin 1 Primary Antibody Unconjugated Public Immunogen Range 824-894/894 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66807 Supplier Bioss Supplier No. BSM90223R
Desmocollin 1 Primary Antibody Unconjugated Public Immunogen Range 824-894/894 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide