Learn More
Bioss KD-Validated DGCR8 Rabbit Reco
SDP
Catalog No. 5000066273
Encompass_Preferred
DGCR8 Microprocessor Complex Subunit Primary Antibody Unconjugated Public Immunogen Range 400-773/773 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66273 Supplier Bioss Supplier No. BSM90280R
DGCR8 Microprocessor Complex Subunit Primary Antibody Unconjugated Public Immunogen Range 400-773/773 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide