Learn More
Bioss KD-Validated Diacylglycerol O-
SDP
Catalog No. 5000066586
Encompass_Preferred
Diacylglycerol O-Acyltransferase 1 Primary Antibody Unconjugated Public Immunogen Range 1-100/488 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66586 Supplier Bioss Supplier No. BSM90218R
Diacylglycerol O-Acyltransferase 1 Primary Antibody Unconjugated Public Immunogen Range 1-100/488 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide