Learn More
Bioss KD-Validated Diaphanous relate
SDP
Catalog No. 5000065127
Encompass_Preferred
Diaphanous Related Formin 1 Primary Antibody Unconjugated Public Immunogen Range 1-45/1272 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65127 Supplier Bioss Supplier No. BSM90008R
Diaphanous Related Formin 1 Primary Antibody Unconjugated Public Immunogen Range 1-45/1272 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide