Learn More
Bioss KD-Validated Dihydrolipoamide
SDP
Catalog No. 5000066849
Encompass_Preferred
Dihydrolipoamide Dehydrogenase Primary Antibody Unconjugated Public Immunogen Range 450-509/509 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66849 Supplier Bioss Supplier No. BSM90104R
Dihydrolipoamide Dehydrogenase Primary Antibody Unconjugated Public Immunogen Range 450-509/509 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide