Learn More
Bioss KD-Validated Dihydropyrimidine
SDP
Catalog No. 5000066280
Encompass_Preferred
Dihydropyrimidine Dehydrogenase Primary Antibody Unconjugated Public Immunogen Range 851-910/1025 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66280 Supplier Bioss Supplier No. BSM90374R
Add to Cart
Add to Cart
Dihydropyrimidine Dehydrogenase Primary Antibody Unconjugated Public Immunogen Range 851-910/1025 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide