Learn More
Bioss KD-Validated DNA ligase 4 Rabb
SDP
Catalog No. 5000066886
Encompass_Preferred
DNA Ligase 4 Primary Antibody Unconjugated Public Immunogen Range 504-685/911 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66886 Supplier Bioss Supplier No. BSM90114R
DNA Ligase 4 Primary Antibody Unconjugated Public Immunogen Range 504-685/911 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide