Learn More
Bioss KD-Validated Drosha ribonuclea
SDP
Catalog No. 5000066179
Encompass_Preferred
Drosha Ribonuclease III Primary Antibody Unconjugated Public Immunogen Range 20-77/1374 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66179 Supplier Bioss Supplier No. BSM90222R
Drosha Ribonuclease III Primary Antibody Unconjugated Public Immunogen Range 20-77/1374 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide