Learn More
Bioss KD-Validated DTYMK Rabbit Reco
SDP
Catalog No. 5000066182
Encompass_Preferred
Deoxythymidylate Kinase Primary Antibody Unconjugated Public Immunogen Range 160-200/212 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66182 Supplier Bioss Supplier No. BSM90226R
Deoxythymidylate Kinase Primary Antibody Unconjugated Public Immunogen Range 160-200/212 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide