Learn More
ABclonal Technology KD Validated DYNC1H1
SDP
Catalog No. 5000010323
Encompass_Preferred
Dyneins are a group of microtubule-activated ATPases that function as molecular motors They are divided into two subgroups of axonemal and cytoplasmic dyneins The cytoplasmic dyneins function in intracellular motility including retrograde axonal transport protein sorting organelle movement and spindle dynamics Molecules of conventional cytoplasmic dynein are comprised of 2 heavy chain polypeptides and a number of intermediate and light chains This gene encodes a member of the cytoplasmic dynein heavy chain family
Catalog No. 50-000-10323 Supplier ABclonal Technology Supplier No. A2729220UL
Dyneins are a group of microtubule-activated ATPases that function as molecular motors They are divided into two subgroups of axonemal and cytoplasmic dyneins The cytoplasmic dyneins function in intracellular motility including retrograde axonal transport protein sorting organelle movement and spindle dynamics Molecules of conventional cytoplasmic dynein are comprised of 2 heavy chain polypeptides and a number of intermediate and light chains This gene encodes a member of the cytoplasmic dynein heavy chain family