Learn More
Bioss KD-Validated DYNLL1 Rabbit Rec
SDP
Catalog No. 5000067035
Encompass_Preferred
Dynein Light Chain LC8-Type 1 Primary Antibody Unconjugated Public Immunogen Range 2450-2511/2511 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67035 Supplier Bioss Supplier No. BSM90418R
Dynein Light Chain LC8-Type 1 Primary Antibody Unconjugated Public Immunogen Range 2450-2511/2511 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide