Learn More
Bioss KD-Validated Early endosome an
SDP
Catalog No. 5000066489
Encompass_Preferred
Early Endosome Antigen 1 Primary Antibody Unconjugated Public Immunogen Range 1-63/1411 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66489 Supplier Bioss Supplier No. BSM90228R
Early Endosome Antigen 1 Primary Antibody Unconjugated Public Immunogen Range 1-63/1411 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide