Learn More
ABclonal Technology KD Validated EIF2AK1
SDP
Catalog No. 5000011270
Encompass_Preferred
The protein encoded by this gene acts at the level of translation initiation to downregulate protein synthesis in response to stress The encoded protein is a kinase that can be inactivated by hemin Two transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-11270 Supplier ABclonal Technology Supplier No. A2461420UL
The protein encoded by this gene acts at the level of translation initiation to downregulate protein synthesis in response to stress The encoded protein is a kinase that can be inactivated by hemin Two transcript variants encoding different isoforms have been found for this gene