Learn More
ABclonal Technology KD Validated EIF2AK1
SDP
Supplier:  ABclonal Technology A24614100UL
Encompass_Preferred
The protein encoded by this gene acts at the level of translation initiation to downregulate protein synthesis in response to stress The encoded protein is a kinase that can be inactivated by hemin Two transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-11269