Learn More
Bioss KD-Validated Ephrin Receptor B
SDP
Catalog No. 5000064984
Encompass_Preferred
EPH Receptor B4 Primary Antibody Unconjugated Public Immunogen Range 5-22/987 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64984 Supplier Bioss Supplier No. BSM90311R
Add to Cart
Add to Cart
EPH Receptor B4 Primary Antibody Unconjugated Public Immunogen Range 5-22/987 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide