Learn More
Bioss KD-Validated Farnesyl diphosph
SDP
Catalog No. 5000066203
Encompass_Preferred
Farnesyl Diphosphate Synthase Primary Antibody Unconjugated Public Immunogen Range 370-410/419 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66203 Supplier Bioss Supplier No. BSM90276R
Farnesyl Diphosphate Synthase Primary Antibody Unconjugated Public Immunogen Range 370-410/419 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide