Learn More
Bioss KD-Validated Fatty acid syntha
SDP
Catalog No. 5000066861
Encompass_Preferred
Fatty Acid Synthase Primary Antibody Unconjugated Public Immunogen Range 100-150/429 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66861 Supplier Bioss Supplier No. BSM90419R
Fatty Acid Synthase Primary Antibody Unconjugated Public Immunogen Range 100-150/429 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide