Learn More
Bioss KD-Validated Forkhead box O4 R
SDP
Catalog No. 5000066274
Encompass_Preferred
Forkhead Box O4 Primary Antibody Unconjugated Public Immunogen Range 450-505/505 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66274 Supplier Bioss Supplier No. BSM90292R
Forkhead Box O4 Primary Antibody Unconjugated Public Immunogen Range 450-505/505 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide