Learn More
Bioss KD-Validated FOXO1 Rabbit Reco
SDP
Catalog No. 5000066941
Encompass_Preferred
Forkhead Box O1 Primary Antibody Unconjugated Public Immunogen Range 450-655/655 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66941 Supplier Bioss Supplier No. BSM90291R
Forkhead Box O1 Primary Antibody Unconjugated Public Immunogen Range 450-655/655 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide