Learn More
Bioss KD-Validated Galactosidase alp
SDP
Catalog No. 5000065481
Encompass_Preferred
Galactosidase Alpha Primary Antibody Unconjugated Public Immunogen Range 10-50/184 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65481 Supplier Bioss Supplier No. BSM90420R
Galactosidase Alpha Primary Antibody Unconjugated Public Immunogen Range 10-50/184 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide