Learn More
Bioss KD-Validated Galactosidase bet
SDP
Catalog No. 5000066850
Encompass_Preferred
Galactosidase Beta 1 Primary Antibody Unconjugated Public Immunogen Range 621-677/677 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66850 Supplier Bioss Supplier No. BSM90285R
Galactosidase Beta 1 Primary Antibody Unconjugated Public Immunogen Range 621-677/677 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide