Learn More
ABclonal Technology KD Validated Gelsolin
SDP
Catalog No. 5000005132
Encompass_Preferred
The protein encoded by this gene binds to the plus ends of actin monomers and filaments to prevent monomer exchange The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments Defects in this gene are a cause of familial amyloidosis Finnish type (FAF) Multiple transcript variants encoding several different isoforms have been found for this gene
Catalog No. 50-000-05132 Supplier ABclonal Technology Supplier No. A2218420UL
The protein encoded by this gene binds to the plus ends of actin monomers and filaments to prevent monomer exchange The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments Defects in this gene are a cause of familial amyloidosis Finnish type (FAF) Multiple transcript variants encoding several different isoforms have been found for this gene