Learn More
ABclonal Technology KD Validated Gelsolin
Supplier: ABclonal Technology A22184100UL
The protein encoded by this gene binds to the plus ends of actin monomers and filaments to prevent monomer exchange The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments Defects in this gene are a cause of familial amyloidosis Finnish type (FAF) Multiple transcript variants encoding several different isoforms have been found for this gene