Learn More
Bioss KD-Validated Glucose-6-phospha
SDP
Catalog No. 5000064982
Encompass_Preferred
Glucose-6-Phosphate Dehydrogenase Primary Antibody Unconjugated Public Immunogen Range 460-515/515 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64982 Supplier Bioss Supplier No. BSM90305R
Glucose-6-Phosphate Dehydrogenase Primary Antibody Unconjugated Public Immunogen Range 460-515/515 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide