Learn More
Bioss KD-Validated Glutamic-oxaloace
SDP
Supplier:  Bioss BSM90422R
Encompass_Preferred
Glutamic-Oxaloacetic Transaminase 1 Primary Antibody Unconjugated Public Immunogen Range 150-203/203 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67034