Learn More
Bioss KD-Validated Glutamic-oxaloace
SDP
Catalog No. 5000064980
Encompass_Preferred
Glutamic-Oxaloacetic Transaminase 2 Primary Antibody Unconjugated Public Immunogen Range 17-72/430 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64980 Supplier Bioss Supplier No. BSM90273R
Glutamic-Oxaloacetic Transaminase 2 Primary Antibody Unconjugated Public Immunogen Range 17-72/430 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide