Learn More
Bioss KD-Validated Glutathione synth
SDP
Catalog No. 5000066169
Encompass_Preferred
Glutathione Synthetase Primary Antibody Unconjugated Public Immunogen Range 321-400/474 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66169 Supplier Bioss Supplier No. BSM90369R
Glutathione Synthetase Primary Antibody Unconjugated Public Immunogen Range 321-400/474 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide