Learn More
Bioss KD-Validated Glycophorin C Rab
SDP
Catalog No. 5000066185
Encompass_Preferred
Glycophorin C Primary Antibody Unconjugated Public Immunogen Range 5-32/196 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66185 Supplier Bioss Supplier No. BSM90393R
Glycophorin C Primary Antibody Unconjugated Public Immunogen Range 5-32/196 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide