Learn More
Bioss KD-Validated Goosecoid homeobo
SDP
Catalog No. 5000066936
Encompass_Preferred
Goosecoid Homeobox Primary Antibody Unconjugated Public Immunogen Range 1-37/257 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66936 Supplier Bioss Supplier No. BSM90265R
Goosecoid Homeobox Primary Antibody Unconjugated Public Immunogen Range 1-37/257 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide