Learn More
Bioss KD-Validated Growth associated
SDP
Catalog No. 5000065433
Encompass_Preferred
Growth Associated Protein 43 Primary Antibody Unconjugated Public Immunogen Range 201-238/238 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65433 Supplier Bioss Supplier No. BSM90298R
Growth Associated Protein 43 Primary Antibody Unconjugated Public Immunogen Range 201-238/238 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide