Learn More
Bioss KD-Validated Growth factor rec
SDP
Supplier:  Bioss BSM90375R
Encompass_Preferred
Growth Factor Receptor Bound Protein 7 Primary Antibody Unconjugated Public Immunogen Range 1-100/532 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66994