Learn More
Bioss KD-Validated Grp75 Rabbit Reco
SDP
Catalog No. 5000066537
Encompass_Preferred
Heat Shock Protein Family A (Hsp70) Member 9 Primary Antibody Unconjugated Public Immunogen Range 410-470/470 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66537 Supplier Bioss Supplier No. BSM90406R
Heat Shock Protein Family A (Hsp70) Member 9 Primary Antibody Unconjugated Public Immunogen Range 410-470/470 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide