Learn More
Bioss KD-Validated HDAC4 Rabbit Reco
SDP
Catalog No. 5000066629
Encompass_Preferred
Histone Deacetylase 4 Primary Antibody Unconjugated Public Immunogen Range 30-100/1084 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66629 Supplier Bioss Supplier No. BSM90300R
Histone Deacetylase 4 Primary Antibody Unconjugated Public Immunogen Range 30-100/1084 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide