Learn More
Bioss KD-Validated HDHD5 Rabbit Reco
SDP
Catalog No. 5000066591
Encompass_Preferred
Haloacid Dehalogenase Like Hydrolase Domain Containing 5 Primary Antibody Unconjugated Public Immunogen Range 370-396/423 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66591 Supplier Bioss Supplier No. BSM90269R
Haloacid Dehalogenase Like Hydrolase Domain Containing 5 Primary Antibody Unconjugated Public Immunogen Range 370-396/423 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide