Learn More
Bioss KD-Validated Hexokinase 1 Rabb
SDP
Catalog No. 5000066990
Encompass_Preferred
Brain Form Hexokinase Primary Antibody Unconjugated Public Immunogen Range 100-156/917 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66990 Supplier Bioss Supplier No. BSM90347R
Brain Form Hexokinase Primary Antibody Unconjugated Public Immunogen Range 100-156/917 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide