Learn More
Bioss KD-Validated Hsp22 Rabbit Reco
SDP
Catalog No. 5000066636
Encompass_Preferred
Heat Shock Protein Family B (Small) Member 8 Primary Antibody Unconjugated Public Immunogen Range 25-48/999 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66636 Supplier Bioss Supplier No. BSM90394R
Heat Shock Protein Family B (Small) Member 8 Primary Antibody Unconjugated Public Immunogen Range 25-48/999 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide