Learn More
ABclonal Technology KD Validated HSP90B1
SDP
Supplier:  ABclonal Technology A2631120UL
Encompass_Preferred
This gene encodes a member of a family of adenosine triphosphate(ATP)-metabolizing molecular chaperones with roles in stabilizing and folding other proteins The encoded protein is localized to melanosomes and the endoplasmic reticulum Expression of this protein is associated with a variety of pathogenic states including tumor formation There is a microRNA gene located within the 5 exon of this gene There are pseudogenes for this gene on chromosomes 1 and 15
Catalog No. 50-000-11728